Assay Details
Gene Symbol: Unc13a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029873.2 | NP_001025044.2 | 17 - 18 | 2180 | 69 | |
GenBank mRNA | AK147408.1 | - | 12 - 13 | 1693 | 69 | |
AY753536.1 | - | 17 - 18 | 2070 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
