Assay Details
Gene Symbol: Ccdc22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138603.3 | NP_613069.3 | 6 - 7 | 841 | 109 | |
GenBank mRNA | AF229637.1 | - | 6 - 7 | 782 | 109 | |
AK010536.1 | - | 6 - 7 | 759 | 109 | ||
AK039573.1 | - | 6 - 7 | 814 | 109 | ||
BC011195.1 | - | 6 - 7 | 746 | 4188459 | 109 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
