Assay Details
Gene Symbol: Cenpi | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK043344.1 | - | 2 - 3 | 198 | 79 | |
AK161473.1 | - | 3 - 4 | 337 | 79 | ||
BC030328.1 | - | 3 - 4 | 254 | 5376020 | 79 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK043344.1, AK161473.1, BC030328.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
