Assay Details
Gene Symbol: AI314180 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172381.2 | NP_759013.2 | 45 - 46 | 5346 | 82 | |
GenBank mRNA | AK030369.1 | - | 5 - 6 | 775 | 82 | |
AK129127.1 | - | 28 - 29 | 3286 | 82 | ||
BC025035.1 | - | 15 - 16 | 1618 | 82 | ||
BC058684.1 | - | 45 - 46 | 5346 | 6834912 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
