Assay Details
Gene Symbol: Herc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025992.2 | NP_080268.1 | 14 - 15 | 1857 | 160 | |
GenBank mRNA | AK011047.1 | - | 14 - 15 | 1744 | 160 | |
AK147960.1 | - | 14 - 15 | 1780 | 160 | ||
AK149648.1 | - | 14 - 15 | 1849 | 160 | ||
AK161293.1 | - | 2 - 3 | 463 | 160 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
