Assay Details
Gene Symbol: Spen | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019763.2 | NP_062737.2 | 12 - 13 | 10300 | 67 | |
GenBank mRNA | AB055980.1 | - | 10 - 11 | 9673 | 67 | |
AF156529.1 | - | 11 - 12 | 9808 | 67 | ||
BC022755.1 | - | 1 - 2 | 736 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
