Assay Details
Gene Symbol: Grin2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008170.2 | NP_032196.2 | 8 - 9 | 2010 | 75 | |
GenBank mRNA | D10217.2 | - | 8 - 9 | 2010 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
