Assay Details
Gene Symbol: Trap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026508.2 | NP_080784.1 | 15 - 16 | 1833 | 75 | |
GenBank mRNA | AK002409.1 | - | 15 - 16 | 1819 | 75 | |
AK010341.1 | - | 15 - 16 | 1833 | 75 | ||
AK088471.1 | - | 15 - 16 | 1822 | 75 | ||
AK143494.1 | - | 15 - 16 | 1851 | 75 | ||
AK162067.1 | - | 15 - 16 | 1821 | 75 | ||
AK167181.1 | - | 15 - 16 | 1822 | 75 | ||
BC006685.1 | - | 15 - 16 | 1801 | 75 | ||
BC006864.1 | - | 15 - 16 | 1801 | 75 | ||
BC022912.1 | - | 15 - 16 | 1830 | 4242394 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
