Assay Details
Gene Symbol: Uba1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001136085.2 | NP_001129557.1 | 8 - 9 | 1041 | 73 | |
NM_001276316.1 | NP_001263245.1 | 9 - 10 | 1133 | 73 | ||
NM_001276317.1 | NP_001263246.1 | 8 - 9 | 1036 | 73 | ||
NM_009457.4 | NP_033483.2 | 8 - 9 | 1036 | 73 | ||
GenBank mRNA | AK081808.1 | - | 8 - 9 | 1007 | 73 | |
AK088057.1 | - | 8 - 9 | 918 | 73 | ||
AK088528.1 | - | 8 - 9 | 918 | 73 | ||
AK143416.1 | - | 8 - 9 | 942 | 73 | ||
AK171667.1 | - | 8 - 9 | 1007 | 73 | ||
AK205753.1 | - | 1 - 2 | 73 | 73 | ||
AK210112.1 | - | 2 - 3 | 223 | 73 | ||
AK219868.1 | - | 1 - 2 | 125 | 73 | ||
AK219938.1 | - | 1 - 2 | 125 | 73 | ||
BC058630.1 | - | 8 - 9 | 982 | 6402368 | 73 | |
BC138200.1 | - | 8 - 9 | 899 | 8861219 | 73 | |
BC145984.1 | - | 8 - 9 | 900 | 40131287 | 73 | |
D10576.1 | - | 8 - 9 | 902 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
