Assay Details
Gene Symbol: Cdk16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011049.4 | NP_035179.1 | 8 - 9 | 1109 | 71 | |
GenBank mRNA | AK051425.1 | - | 8 - 9 | 1110 | 71 | |
AK141083.1 | - | 2 - 3 | 128 | 71 | ||
AK143518.1 | - | 8 - 9 | 1137 | 71 | ||
AK145483.1 | - | 8 - 9 | 1139 | 71 | ||
AK166188.1 | - | 8 - 9 | 1318 | 71 | ||
AK179102.1 | - | 2 - 3 | 162 | 71 | ||
AK180999.1 | - | 2 - 3 | 162 | 71 | ||
AK194254.1 | - | 2 - 3 | 162 | 71 | ||
AK203854.1 | - | 2 - 3 | 162 | 71 | ||
AK214881.1 | - | 3 - 4 | 225 | 71 | ||
BC011069.1 | - | 8 - 9 | 852 | 4006745 | 71 | |
BC013663.1 | - | 8 - 9 | 1168 | 4208974 | 71 | |
X69025.1 | - | 8 - 9 | 1131 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
