Assay Details
Gene Symbol: Zc3h7a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145931.2 | NP_666043.2 | 2 - 3 | 276 | 98 | |
NR_027502.1 | 2 - 3 | 276 | 98 | |||
GenBank mRNA | AK028438.1 | - | 2 - 3 | 200 | 99 | |
AK040507.1 | - | 2 - 3 | 224 | 98 | ||
AK049834.1 | - | 2 - 3 | 211 | 98 | ||
AK052830.1 | - | 3 - 4 | 387 | 98 | ||
AK150952.1 | - | 2 - 3 | 195 | 98 | ||
AK161148.1 | - | 2 - 3 | 254 | 98 | ||
AK166876.1 | - | 2 - 3 | 224 | 98 | ||
AK167758.1 | - | 2 - 3 | 225 | 98 | ||
BC027330.1 | - | 2 - 3 | 172 | 3674968 | 98 | |
BC029320.1 | - | 2 - 3 | 174 | 98 | ||
BY297607.1 | - | 2 - 3 | 277 | 98 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AK028438.1, AK166876.1, BC027330.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
