Assay Details
Gene Symbol: Fgd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF402611.1 | - | 6 - 7 | 1158 | 78 | |
AF402612.1 | - | 5 - 6 | 1055 | 78 | ||
AF402613.1 | - | 6 - 7 | 1205 | 78 | ||
AK028302.1 | - | 3 - 4 | 607 | 78 | ||
AK034504.1 | - | 5 - 6 | 1148 | 78 | ||
AK043656.1 | - | 6 - 7 | 1203 | 78 | ||
AK054242.1 | - | 6 - 7 | 1146 | 78 | ||
AK134961.1 | - | 6 - 7 | 1221 | 78 | ||
BC129902.1 | - | 6 - 7 | 1245 | 40111637 | 78 | |
BC129903.1 | - | 6 - 7 | 1246 | 40111639 | 78 | |
BC145523.1 | - | 6 - 7 | 1299 | 78 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
