Assay Details
Gene Symbol: Dnm1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025947.2 | NP_001021118.1 | 5 - 6 | 622 | 71 | |
NM_001276340.1 | NP_001263269.1 | 5 - 6 | 640 | 71 | ||
NM_001276341.1 | NP_001263270.1 | 4 - 5 | 384 | 71 | ||
NM_152816.3 | NP_690029.2 | 6 - 7 | 661 | 71 | ||
NR_075074.1 | 5 - 6 | 431 | 71 | |||
GenBank mRNA | AK018195.1 | - | 6 - 7 | 601 | 71 | |
AK051443.1 | - | 5 - 6 | 551 | 71 | ||
AK080871.1 | - | 5 - 6 | 542 | 71 | ||
AK154123.1 | - | 5 - 6 | 391 | 71 | ||
BB612294.1 | - | 6 - 7 | 489 | 71 | ||
BC027538.1 | - | 5 - 6 | 542 | 71 | ||
BC040777.1 | - | 4 - 5 | 354 | 4948368 | 71 | |
BC079635.1 | - | 5 - 6 | 471 | 6400313 | 71 | |
CD561569.1 | - | 4 - 5 | 348 | 71 | ||
CK634757.1 | - | 3 - 4 | 193 | 71 | ||
CO432650.1 | - | 2 - 3 | 150 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
