Assay Details
Gene Symbol: Rnf20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163263.1 | NP_001156735.1 | 9 - 10 | 1575 | 121 | |
NM_182999.2 | NP_892044.1 | 10 - 11 | 1422 | 121 | ||
GenBank mRNA | AK048862.1 | - | 9 - 10 | 1534 | 121 | |
AK076501.1 | - | 5 - 6 | 504 | 121 | ||
AK139883.1 | - | 7 - 8 | 950 | 121 | ||
AK163905.1 | - | 6 - 7 | 787 | 121 | ||
AK220492.1 | - | 9 - 10 | 1491 | 121 | ||
BC004593.1 | - | 5 - 6 | 649 | 121 | ||
BC052482.1 | - | 10 - 11 | 1361 | 6308280 | 121 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
