Assay Details
Gene Symbol: Phldb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252442.1 | NP_001239371.1 | 9 - 10 | 2656 | 131 | |
NM_153412.4 | NP_700461.2 | 9 - 10 | 2656 | 131 | ||
GenBank mRNA | AK050493.1 | - | 10 - 11 | 2823 | 131 | |
AK202029.1 | - | 3 - 4 | 218 | 131 | ||
BC050915.1 | - | 9 - 10 | 2512 | 6405379 | 131 | |
BC060683.1 | - | 9 - 10 | 2512 | 6405379 | 131 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
