Assay Details
Gene Symbol: Usp16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024258.2 | NP_077220.2 | 15 - 16 | 2269 | 144 | |
GenBank mRNA | AK004594.2 | - | 15 - 16 | 2244 | 144 | |
AK013437.1 | - | 9 - 10 | 1285 | 144 | ||
AK160038.1 | - | 15 - 16 | 2239 | 144 | ||
BC003278.1 | - | 15 - 16 | 2198 | 3582180 | 144 | |
BC004577.1 | - | 3 - 4 | 781 | 144 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
