Assay Details
Gene Symbol: Usp16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024258.2 | NP_077220.2 | 16 - 17 | 2356 | 139 | |
GenBank mRNA | AK004594.2 | - | 16 - 17 | 2331 | 139 | |
AK013437.1 | - | 10 - 11 | 1372 | 139 | ||
AK160038.1 | - | 16 - 17 | 2326 | 139 | ||
BC003278.1 | - | 16 - 17 | 2285 | 3582180 | 139 | |
BC004577.1 | - | 4 - 5 | 868 | 139 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
