Assay Details
Gene Symbol: Slc4a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001136260.1 | NP_001129732.1 | 22 - 23 | 3004 | 98 | |
NM_001197147.1 | NP_001184076.1 | 22 - 23 | 2977 | 98 | ||
NM_018760.2 | NP_061230.2 | 22 - 23 | 3004 | 98 | ||
GenBank mRNA | AF141934.3 | - | 19 - 20 | 2806 | 98 | |
AW124701.1 | - | 1 - 2 | 98 | |||
BC026592.1 | - | 13 - 14 | 1797 | 98 | ||
CF744823.1 | - | 1 - 2 | 132 | 98 | ||
HQ018820.1 | - | 19 - 20 | 2841 | 98 | ||
HQ204220.1 | - | 22 - 23 | 2948 | 98 | ||
HQ285250.1 | - | 19 - 20 | 2817 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
