Assay Details
Gene Symbol: Oxsr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133985.2 | NP_598746.2 | 10 - 11 | 1311 | 87 | |
NR_045693.1 | 11 - 12 | 1440 | 87 | |||
GenBank mRNA | AK031790.1 | - | 9 - 10 | 808 | 87 | |
AK075837.1 | - | 9 - 10 | 819 | 87 | ||
AK129291.1 | - | 10 - 11 | 1306 | 87 | ||
BC036148.1 | - | 1 - 2 | 61 | 87 | ||
BC046453.1 | - | 11 - 12 | 1335 | 87 | ||
BC060645.1 | - | 10 - 11 | 1294 | 6852021 | 87 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
