Assay Details
Gene Symbol: Lpcat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK046079.1 | - | 1 - 2 | 287 | 67 | |
AK046507.1 | - | 1 - 2 | 287 | 67 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK046079.1, AK046507.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
