Assay Details
Gene Symbol: Golgb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030035.1 | NP_084311.1 | 9 - 10 | 1477 | 136 | |
GenBank mRNA | BC145221.1 | - | 9 - 10 | 1436 | 9053772 | 136 |
BC150731.1 | - | 9 - 10 | 1454 | 9087642 | 136 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
