Assay Details
Gene Symbol: Ctnnb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001165902.1 | NP_001159374.1 | 11 - 12 | 1887 | 80 | |
NM_007614.3 | NP_031640.1 | 11 - 12 | 2087 | 80 | ||
GenBank mRNA | AK018515.1 | - | 11 - 12 | 2001 | 80 | |
AK035311.1 | - | 11 - 12 | 2000 | 80 | ||
AK132398.1 | - | 11 - 12 | 2068 | 80 | ||
AK133656.1 | - | 11 - 12 | 2000 | 80 | ||
AK135743.1 | - | 8 - 9 | 1554 | 80 | ||
AK189807.1 | - | 1 - 2 | 42 | 80 | ||
AK190684.1 | - | 1 - 2 | 42 | 80 | ||
AY751549.1 | - | 11 - 12 | 2014 | 80 | ||
BC043078.1 | - | 8 - 9 | 1489 | 80 | ||
BC043481.1 | - | 7 - 8 | 1121 | 80 | ||
BC048153.1 | - | 11 - 12 | 2012 | 5709247 | 80 | |
BC053065.1 | - | 11 - 12 | 2012 | 5709247 | 80 | |
M90364.1 | - | 11 - 12 | 1897 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
