Assay Details
Gene Symbol: Gm19619 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_040428.1 | 1 - 2 | 84 | 66 | ||
GenBank mRNA | AK050091.1 | - | 1 - 2 | 85 | 66 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | amphiregulin | Areg |
AR, Mcub, Sdgf |
-133.0kb | Anti-sense | Chr. 5 - 91139615 - 91148432 on NCBI Build GRCm38 |
Downstream |
betacellulin, epidermal growth factor family member |
Btc | Bcn | 74.2kb | Sense | Chr. 5 - 91357261 - 91402994 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC1400000269.rn.1
Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
