Assay Details
Gene Symbol: Gm30790 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK038693.1 | - | 2 - 3 | 289 | 70 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | nuclear receptor interacting protein 1 | Nrip1 |
6030458L20Rik, 8430438I05Rik, 9630050P12, AA959574, AW456757, RIP140 |
-378.7kb | Sense | Chr. 16 - 76287403 - 76374195 on NCBI Build GRCm38 |
Downstream | RIKEN cDNA 1700041M19 gene | 1700041M19Rik | 249.9kb | Sense | Chr. 16 - 77004228 - 77010881 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
