Assay Details
Gene Symbol: Pde9a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163748.1 | NP_001157220.1 | 4 - 5 | 310 | 58 | |
NM_008804.4 | NP_032830.3 | 4 - 5 | 310 | 58 | ||
GenBank mRNA | AF031147.1 | - | 4 - 5 | 265 | 58 | |
AF068247.1 | - | 4 - 5 | 274 | 58 | ||
AK030509.1 | - | 4 - 5 | 294 | 58 | ||
BC025882.1 | - | 4 - 5 | 268 | 58 | ||
BC038675.1 | - | 4 - 5 | 259 | 58 | ||
BC046915.1 | - | 4 - 5 | 259 | 58 | ||
BC061163.1 | - | 4 - 5 | 290 | 6533665 | 58 | |
BY283127.1 | - | 4 - 5 | 311 | 58 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
