Assay Details
Gene Symbol: Npr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173788.3 | NP_776149.1 | 16 - 17 | 2761 | 89 | |
GenBank mRNA | AK158195.1 | - | 11 - 12 | 1312 | 89 | |
BC023420.1 | - | 3 - 4 | 402 | 89 | ||
BC037458.1 | - | 11 - 12 | 1217 | 89 | ||
BC042470.1 | - | 16 - 17 | 2761 | 5137261 | 89 | |
BC049896.1 | - | 6 - 7 | 856 | 89 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
