Assay Details
Gene Symbol: Pik3r2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK146195.1 | - | 1 - 2 | 99 | 109 | |
AK148122.1 | - | 1 - 2 | 106 | 109 | ||
BC006796.1 | - | 1 - 2 | 100 | 2581969 | 109 | |
Y13569.1 | - | 1 - 2 | 78 | 109 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK146195.1, BC006796.1, Y13569.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
MicroArray Probes:
