Assay Details
Gene Symbol: Dsp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023842.2 | NP_076331.2 | 17 - 18 | 2761 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
