Assay Details
Gene Symbol: Nup153 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175749.2 | NP_786925.2 | 18 - 19 | 3889 | 127 | |
GenBank mRNA | AK031006.1 | - | 1 - 2 | 620 | 127 | |
BC027402.1 | - | 1 - 2 | 521 | 127 | ||
BC046468.1 | - | 6 - 7 | 1913 | 127 | ||
BC052173.1 | - | 18 - 19 | 3627 | 6516328 | 127 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
