Assay Details
Gene Symbol: Tll1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009390.2 | NP_033416.2 | 18 - 19 | 3045 | 122 | |
GenBank mRNA | AK139103.1 | - | 6 - 7 | 834 | 122 | |
U34042.1 | - | 18 - 19 | 3045 | 122 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
