Assay Details
Gene Symbol: Slc12a7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011390.2 | NP_035520.1 | 15 - 16 | 2033 | 77 | |
GenBank mRNA | AF087436.1 | - | 15 - 16 | 2035 | 77 | |
AK131129.1 | - | 15 - 16 | 1987 | 77 | ||
AK143535.1 | - | 15 - 16 | 2027 | 77 | ||
AK149750.1 | - | 15 - 16 | 2028 | 77 | ||
AK166215.1 | - | 15 - 16 | 2027 | 77 | ||
AK169950.1 | - | 15 - 16 | 2023 | 77 | ||
AK171498.1 | - | 15 - 16 | 2070 | 77 | ||
BC059242.1 | - | 15 - 16 | 2025 | 77 | ||
BC060133.1 | - | 15 - 16 | 2025 | 77 | ||
BC141107.1 | - | 15 - 16 | 1993 | 9055719 | 77 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
