Assay Details
Gene Symbol: Ahrr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009644.2 | NP_033774.1 | 1 - 2 | 82 | 65 | |
GenBank mRNA | AK154152.1 | - | 1 - 2 | 71 | 65 | |
AK155711.1 | - | 1 - 2 | 104 | 65 | ||
BC118534.1 | - | 1 - 2 | 70 | 40095111 | 65 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK154152.1, AK155711.1, BC118534.1, NM_009644.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
