Assay Details
Gene Symbol: Ahrr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009644.2 | NP_033774.1 | 3 - 4 | 335 | 76 | |
GenBank mRNA | AB015140.1 | - | 3 - 4 | 335 | 76 | |
AK154152.1 | - | 3 - 4 | 324 | 76 | ||
AK154761.1 | - | 3 - 4 | 357 | 76 | ||
AK155711.1 | - | 3 - 4 | 357 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
