Assay Details
Gene Symbol: Exoc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177333.3 | NP_796307.2 | 1 - 2 | 43 | 64 | |
GenBank mRNA | AK088333.1 | - | 1 - 2 | 37 | 64 | |
AK141537.1 | - | 1 - 2 | 43 | 64 | ||
BC031743.1 | - | 1 - 2 | 31 | 4187527 | 64 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AK088333.1, AK141537.1, BC031743.1, NM_177333.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
