Assay Details
Gene Symbol: Exoc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177333.3 | NP_796307.2 | 5 - 6 | 1255 | 79 | |
GenBank mRNA | AK088333.1 | - | 5 - 6 | 1249 | 79 | |
AK089979.1 | - | 5 - 6 | 1265 | 79 | ||
AK131141.1 | - | 5 - 6 | 1239 | 79 | ||
AK141537.1 | - | 5 - 6 | 1255 | 79 | ||
BC031743.1 | - | 5 - 6 | 1243 | 4187527 | 79 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
