Assay Details
Gene Symbol: Usp51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001137547.1 | NP_001131019.1 | 1 - 2 | 79 | 84 | |
GenBank mRNA | AK132077.1 | - | 1 - 2 | 79 | 84 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the NM_001137547.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
Function(s)
Process(es)
regulation of transcription, DNA-templated
histone deubiquitination
protein deubiquitination
proteasome-mediated ubiquitin-dependent protein catabolic process
regulation of proteasomal protein catabolic process
cysteine-type endopeptidase activity
thiol-dependent deubiquitinase
regulation of transcription, DNA-templated
histone deubiquitination
protein deubiquitination
proteasome-mediated ubiquitin-dependent protein catabolic process
regulation of proteasomal protein catabolic process
cysteine-type endopeptidase activity
thiol-dependent deubiquitinase
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1959218 | NM_001137547.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2176953 | NM_001137547.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 834307 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2176953 | NM_001137547.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1959218 | NM_001137547.1 | mouse_transcriptome |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32751537 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32751560 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14812543 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32751531 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14829377 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32751585 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32751571 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32751559 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32751558 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32751551 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32751578 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32751555 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32751567 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32751581 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32751565 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32751570 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32751563 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32751543 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14618225 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32751575 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32751583 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32751562 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32751545 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32751527 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32751534 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32751568 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32751572 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32751587 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32751487 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32751557 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32751569 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14642232 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32751475 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32751525 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32751554 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32751538 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32751540 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14812544 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32751529 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32751577 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32751548 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32751566 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32751526 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10602689 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10602688 | - | - |
Affymetrix Mouse Clariom S Array | PSR0X00009872.mm.1 | TC0X00001551.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0X00009872.mm.1 | NM_001137547.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0X00009872.mm.1 | NM_001137547.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0X00009873.mm.1 | TC0X00001551.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0X00009873.mm.1 | NM_001137547.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0X00009873.mm.1 | NM_001137547.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0X00009874.mm.1 | TC0X00001551.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0X00009874.mm.1 | NM_001137547.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0X00009874.mm.1 | NM_001137547.1 | Refseq |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 456721 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.1 expression beadchip | ILMN_1244361 | - | - |
