Assay Details
Gene Symbol: Secisbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK008918.1 | - | 2 - 3 | 324 | 71 | |
AK030021.1 | - | 11 - 12 | 1618 | 71 | ||
AK156076.1 | - | 11 - 12 | 1694 | 71 | ||
AK168634.1 | - | 11 - 12 | 1651 | 71 | ||
BC125391.1 | - | 11 - 12 | 1674 | 40129906 | 71 | |
BC125393.1 | - | 11 - 12 | 1671 | 40129908 | 71 | |
BC144795.1 | - | 11 - 12 | 1669 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
