Assay Details
Gene Symbol: Secisbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK008918.1 | - | 3 - 4 | 466 | 87 | |
AK030021.1 | - | 12 - 13 | 1760 | 87 | ||
AK156076.1 | - | 12 - 13 | 1836 | 87 | ||
AK168634.1 | - | 12 - 13 | 1793 | 87 | ||
BC125391.1 | - | 12 - 13 | 1816 | 40129906 | 87 | |
BC125393.1 | - | 12 - 13 | 1813 | 40129908 | 87 | |
BC144795.1 | - | 12 - 13 | 1811 | 87 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
