Assay Details
Gene Symbol: Ngef | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111314.1 | NP_001104784.1 | 12 - 13 | 2009 | 61 | |
NM_019867.2 | NP_063920.2 | 10 - 11 | 1646 | 61 | ||
GenBank mRNA | AJ238898.1 | - | 10 - 11 | 1627 | 61 | |
AK138433.1 | - | 10 - 11 | 1615 | 61 | ||
AY038025.1 | - | 10 - 11 | 1621 | 61 | ||
BC022680.1 | - | 4 - 5 | 424 | 61 | ||
BC039279.1 | - | 12 - 13 | 2009 | 4973826 | 61 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC022680.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
