Assay Details
Gene Symbol: Kbtbd8os | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK005835.1 | - | 1 - 2 | 44 | 62 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | RIKEN cDNA 4930511E03 gene | 4930511E03Rik | -162.5kb | Anti-sense | Chr. 6 - 94943834 - 94951545 on NCBI Build GRCm38 | |
Downstream | kelch repeat and BTB (POZ) domain containing 8 | Kbtbd8 |
AU040152, SSEC-51, SSEC51, Takrp, mKIAA1842 |
249.0 | Anti-sense | Chr. 6 - 95117880 - 95129793 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
