Assay Details
Gene Symbol: Bag5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027404.2 | NP_081680.1 | 2 | 1047 | 115 | |
GenBank mRNA | AK005534.1 | - | 1 | 864 | 115 | |
AK160246.1 | - | 2 | 1169 | 115 | ||
BC023099.1 | - | 2 | 971 | 115 | ||
BC027827.1 | - | 1 | 83 | 115 | ||
BC037642.1 | - | 2 | 1059 | 5066679 | 115 | |
BC055762.1 | - | 2 | 967 | 115 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of protein ubiquitination
negative regulation of proteasomal ubiquitin-dependent protein catabolic process
negative regulation of ubiquitin-protein transferase activity
negative regulation of protein refolding
neuron death
regulation of inclusion body assembly
negative regulation of oxidative stress-induced intrinsic apoptotic signaling pathway
protein binding
protein kinase binding
ubiquitin protein ligase binding
chaperone binding
negative regulation of protein ubiquitination
negative regulation of proteasomal ubiquitin-dependent protein catabolic process
negative regulation of ubiquitin-protein transferase activity
negative regulation of protein refolding
neuron death
regulation of inclusion body assembly
negative regulation of oxidative stress-induced intrinsic apoptotic signaling pathway
protein binding
protein kinase binding
ubiquitin protein ligase binding
chaperone binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28745653 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28745652 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28745650 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28745651 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2003648 | BC037642.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2003648 | BC023099.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2003648 | BC055762.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2003648 | NM_027404.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2003648 | AK005534.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 90034 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P119081 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1999419 | BC023099.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1999419 | BC055762.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1999419 | NM_027404.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2003648 | BC037642.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2003648 | BC023099.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2003648 | BC055762.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2003648 | NM_027404.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2003648 | AK005534.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P216315 | BC027827.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P216315 | BC037642.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P216315 | BC023099.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P216315 | BC055762.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P216315 | NM_027404.2 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28745638 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28745645 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28745626 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28745629 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28745646 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28745616 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28745619 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28745618 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28745613 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28745623 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28745615 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28745648 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28745644 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28745640 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28745642 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28745652 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28745634 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28745627 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28745621 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28745628 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28745636 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28745633 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28745625 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28745632 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28745631 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28745650 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P119081 | - | - |
Whole Mouse Genome G4122A | A_51_P216315 | BC027827.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P216315 | BC037642.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P216315 | BC023099.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P216315 | BC055762.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P216315 | NM_027404.2 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P119081 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28745652 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28745650 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28745653 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28745651 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28745593 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28745642 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28745614 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28745638 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28745598 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28745608 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28745600 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28745634 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28745562 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28745628 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28745568 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28745591 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28745648 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28745631 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28745652 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28745574 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28745621 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28745589 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28745611 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28745592 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28745585 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28745629 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28745577 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28745616 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28745646 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28745650 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28745618 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28745583 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28745571 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28745640 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28745597 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28745606 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28745604 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28745626 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28745636 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28745564 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28745582 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28745623 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28745644 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28745645 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28745601 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28745625 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28745573 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28745588 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28745619 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28745612 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28745606 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10156608 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10156613 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P09865803 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10156618 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10389496 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28745603 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28745629 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10113544 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10156607 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28745646 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28745638 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10156604 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28745625 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28745618 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28745608 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28745611 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28745623 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28745636 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28745631 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28745616 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28745600 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28745628 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28745614 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28745604 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28745645 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10389499 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28745612 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10156596 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28745619 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28745626 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10402720 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10402719 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10402718 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10402717 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10402715 | - | - |
Mouse Genome 430 2.0 Array | 1432638_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1432638_at | - | - |
Mouse Genome 430 2.0 Array | 1452628_at | - | - |
Mouse Genome 430 2.0 Array | 1452628_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 70369_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 70369_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1200016748.mm.1 | TC1200002413.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1200016748.mm.1 | NM_027404.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1200016748.mm.1 | NM_027404.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1200016749.mm.1 | TC1200002413.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1200016749.mm.1 | NM_027404.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1200016749.mm.1 | NM_027404.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1200016752.mm.1 | TC1200002413.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1200016752.mm.1 | NM_027404.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1200016752.mm.1 | NM_027404.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1200016750.mm.1 | TC1200002413.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1200016750.mm.1 | NM_027404.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1200016750.mm.1 | NM_027404.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1200016758.mm.1 | TC1200002413.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1200016758.mm.1 | NM_027404.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1200016758.mm.1 | NM_027404.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1200016751.mm.1 | TC1200002413.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1200016751.mm.1 | NM_027404.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1200016751.mm.1 | NM_027404.2 | Refseq |
Mouse Expression Set 430 | 1452628_PM_at | - | - |
Mouse Expression Set 430 | 1452628_at | - | - |
Mouse Expression Set 430 | 1432638_at | - | - |
Mouse Expression Set 430 | 1432638_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1452628_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1452628_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 775568 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_3163192 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_3163192 | - | - |
mouse-6 v1.1 (Illumina) | GI_58037204-A | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_3106819 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3106819 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | GI_58037204-A | - | - |
