Assay Details
Gene Symbol: Gvin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039160.2 | NP_001034249.2 | 2 | 8463 | 81 | |
NM_029000.3 | NP_083276.2 | 4 | 8657 | 81 | ||
GenBank mRNA | AK151973.1 | - | 1 | 845 | 81 | |
AK153479.1 | - | 1 | 845 | 81 |
Gene Symbol: Gm4070 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243039.1 | NP_001229968.1 | 4 | 8654 | 81 | |
NM_001243040.1 | NP_001229969.1 | 2 | 8460 | 81 | ||
GenBank mRNA | AY167972.1 | - | 4 | 8643 | 81 | |
AY167973.1 | - | 2 | 8432 | 81 | ||
AY167974.1 | - | 4 | 8626 | 81 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P535484 | AY167972.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P535484 | AY167973.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P535484 | AY167974.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P535484 | NM_001039160.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P535484 | NM_029000.3 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P535484 | AY167972.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P535484 | AY167973.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P535484 | AY167974.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P535484 | NM_001039160.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P535484 | NM_029000.3 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1996973 | AK151973.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1996973 | AK153479.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1996973 | AY167972.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1996973 | AY167973.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1996973 | AY167974.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P138367 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P144103 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P535484 | AY167972.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P535484 | AY167973.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P535484 | AY167974.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P535484 | NM_001039160.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P535484 | NM_029000.3 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P535484 | AY167972.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P535484 | AY167973.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P535484 | AY167974.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P535484 | NM_001039160.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P535484 | NM_029000.3 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1996973 | AK151973.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1996973 | AK153479.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1996973 | AY167972.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1996973 | AY167973.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1996973 | AY167974.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25395196 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25395188 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P25395178 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P144103 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P138367 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06312183 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06312161 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06312185 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06312231 | - | - |
Whole Mouse Genome G4122A | A_52_P535484 | AY167972.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P535484 | AY167973.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P535484 | AY167974.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P535484 | NM_001039160.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P535484 | NM_029000.3 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P535484 | AY167972.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P535484 | AY167973.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P535484 | AY167974.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P535484 | NM_001039160.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P535484 | NM_029000.3 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P251690 | AK018585.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P251690 | NR_030720.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P251690 | BC030868.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P251690 | NR_030719.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P251690 | AK151973.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P661713 | AK151973.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P661713 | AK153479.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P661713 | AY167972.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P661713 | NM_001039160.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P661713 | NM_029000.3 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P105299 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P122566 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P125860 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P138123 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P144103 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P138367 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P06312183 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25395086 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25395133 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25395092 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25395064 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25395138 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25395161 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25395078 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25395103 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25395081 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25395110 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25395196 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25395074 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25395139 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25395129 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25395144 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25395057 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25395118 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25395080 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25395065 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25395153 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25395164 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25395073 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25395162 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25395148 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25395168 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25395156 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25395113 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25395076 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25395068 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25395136 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25395079 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25395075 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25395120 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25395166 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06579913 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25395170 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25395086 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06326863 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25395164 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25395168 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25395095 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25395092 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25395121 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25395181 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25395103 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25395162 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25395110 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25395178 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25395196 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06326834 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25395166 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06406506 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25395120 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25395188 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10566575 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10566577 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10566577 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10566576 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10566573 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10566573 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10566575 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10566579 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10566576 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10566579 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1429184_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10566574 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10566571 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10566571 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10566578 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10566578 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10566574 | - | - |
Mouse Genome 430 2.0 Array | 1429184_at | - | - |
Mouse Genome 430 2.0 Array | 1429184_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0700033356.mm.1 | TC0700003996.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0700033356.mm.1 | NM_001243039.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700033356.mm.1 | NM_001243039.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700033356.mm.1 | NM_001243040.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700033356.mm.1 | NM_001243040.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700033356.mm.1 | NM_029000.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700033360.mm.1 | TC0700003996.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0700033360.mm.1 | NM_001243039.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700033360.mm.1 | NM_001243039.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700033360.mm.1 | NM_029000.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700033361.mm.1 | TC0700003996.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0700033361.mm.1 | NM_001243039.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700033361.mm.1 | NM_001243039.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700033361.mm.1 | NM_029000.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700033385.mm.1 | TC0700004002.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0700033385.mm.1 | NM_001039160.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700033385.mm.1 | NM_001039160.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0700033385.mm.1 | NM_029000.3.1 | Refseq |
Mouse Expression Set 430 | 1429184_PM_at | - | - |
Mouse Expression Set 430 | 1429184_PM_at | - | - |
Mouse Expression Set 430 | 1429184_at | - | - |
Mouse Expression Set 430 | 1429184_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 923384 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 4670132 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 4670132 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 3190110 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0004670132 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0003190110 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2739905 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1248351 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2739905 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1248351 | - | - |
mouse-6 v1.1 (Illumina) | scl0074558.2_172-S | - | - |
mouse-6 v1.1 (Illumina) | scl074558.8_66-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 3190110 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 4670132 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl0074558.2_172-S | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl074558.8_66-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4670132 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3190110 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2739905 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1248351 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_3120510 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1248351 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2739905 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3120510 | - | - |
