Assay Details
Gene Symbol: Slc47a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033542.2 | NP_001028714.1 | 6 - 7 | 653 | 67 | |
GenBank mRNA | AK133432.1 | - | 6 - 7 | 654 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
