Assay Details
Gene Symbol: Gpr171 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173398.3 | NP_775574.1 | 2 | 1632 | 125 | |
GenBank mRNA | AK041381.1 | - | 2 | 1604 | 125 | |
AK041582.1 | - | 2 | 1628 | 125 | ||
AK083323.1 | - | 2 | 1630 | 125 | ||
AK089252.1 | - | 2 | 1627 | 125 | ||
AK089287.1 | - | 2 | 1585 | 125 | ||
AK154725.1 | - | 2 | 1630 | 125 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P473498 | AK154725.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P473498 | AK089252.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P473498 | AK083323.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P473498 | AK089287.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P473498 | NM_173398.3 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P473498 | AK154725.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P473498 | AK089252.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P473498 | AK083323.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P473498 | AK089287.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P473498 | NM_173398.3 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P162921 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P473498 | AK154725.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P473498 | AK089252.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P473498 | AK083323.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P473498 | AK089287.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P473498 | NM_173398.3 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P473498 | AK154725.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P473498 | AK089252.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P473498 | AK083323.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P473498 | AK089287.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P473498 | NM_173398.3 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P473498 | AK154725.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P473498 | AK089252.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P473498 | AK083323.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P473498 | AK089287.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P473498 | NM_173398.3 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P473498 | AK154725.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P473498 | AK089252.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P473498 | AK083323.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P473498 | AK089287.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P473498 | NM_173398.3 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22171948 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22171964 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22171955 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22171971 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22171940 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22171981 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22171975 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22171967 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22171977 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22171958 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22171945 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22171983 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22171979 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22171982 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22171949 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22171947 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22171965 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22171973 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22171953 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22171942 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22171950 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22171966 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22171951 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22171959 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22171960 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22171946 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P162921 | - | - |
Whole Mouse Genome G4122A | A_51_P473498 | AK154725.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P473498 | AK089252.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P473498 | AK083323.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P473498 | AK089287.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P473498 | NM_173398.3 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P473498 | AK154725.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P473498 | AK089252.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P473498 | AK083323.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P473498 | AK089287.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P473498 | NM_173398.3 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P115641 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P162921 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02383198 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02383201 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02526922 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02383184 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22171923 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22171933 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22171921 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22171880 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22171925 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22171887 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22171871 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22171959 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22171927 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22171962 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22171945 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22171865 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22171946 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22171875 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22171888 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22171912 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22171938 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22171991 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22171928 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22171935 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22171870 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22171868 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22171896 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22171885 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22171965 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22171940 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22171902 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22171873 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22171922 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22171917 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22171953 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22171942 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22171929 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22171958 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22171973 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22171904 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22171898 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22171909 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22171907 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22171951 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22171882 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22171879 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22171981 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22171979 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22171949 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22171872 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22171931 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22171876 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22171950 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22171987 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22171967 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22171862 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22171955 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22171869 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22171936 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22171914 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22171971 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22171985 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22171918 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22171889 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22171890 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22171915 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22171895 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22171978 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22171901 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22171975 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22171988 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22171925 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22171987 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22171890 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22171915 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22171938 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02732133 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22171962 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22171891 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22171977 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22171896 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22171888 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22171901 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22171945 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22171919 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22171958 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22171949 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22171914 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22171981 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22171960 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22171951 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02562320 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22171936 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02868259 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22171959 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22171940 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22171904 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22171889 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22171920 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22171964 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02835701 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22171953 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22171916 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03197553 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02732140 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22171922 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02589642 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22171935 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22171929 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22171973 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22171931 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22171985 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22171946 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02732135 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22171894 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22171967 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22171955 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02732136 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22171912 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22171927 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22171902 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22171976 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22171950 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22171948 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22171898 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22171983 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22171965 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22171917 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22171947 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22171907 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22171918 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22171942 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22171971 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22171895 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22171910 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10498347 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10498346 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10498348 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1438439_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10498345 | - | - |
Mouse Genome 430 2.0 Array | 1438439_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1438439_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 229323_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 229323_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0300015480.mm.1 | NM_173398.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300015480.mm.1 | NM_173398.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300015480.mm.1 | TC0300002050.mm.1 | MTA TC |
Mouse Expression Set 430 | 1438439_PM_at | - | - |
Mouse Expression Set 430 | 1438439_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1438439_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1438439_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 6760035 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0006760035 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1256824 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1256824 | - | - |
mouse-6 v1.1 (Illumina) | scl22124.2_66-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 6760035 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl22124.2_66-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6760035 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1256824 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1256824 | - | - |
