Assay Details
Gene Symbol: Akap4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042542.2 | NP_001036007.1 | 5 | 2446 | 113 | |
NM_009651.4 | NP_033781.2 | 5 | 2442 | 113 | ||
GenBank mRNA | BC130271.1 | - | 5 | 2374 | 40143193 | 113 |
U07423.1 | - | 5 | 2415 | 113 | ||
U10341.1 | - | 5 | 2447 | 113 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
transmembrane receptor protein serine/threonine kinase signaling pathway
protein localization
regulation of protein kinase A signaling
cell projection organization
sperm motility
motile cilium assembly
establishment of protein localization
protein binding
protein kinase A binding
transmembrane receptor protein serine/threonine kinase signaling pathway
protein localization
regulation of protein kinase A signaling
cell projection organization
sperm motility
motile cilium assembly
establishment of protein localization
protein binding
protein kinase A binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2041976 | U10341.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2041976 | NM_009651.3 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2041976 | NM_001042542.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2110052 | U10341.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2110052 | NM_009651.3 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2110052 | NM_001042542.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 123040 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 537645 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 644216 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 106520 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 548846 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 172738 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P157795 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P110585 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2110052 | U10341.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2110052 | NM_009651.3 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2110052 | NM_001042542.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2041976 | U10341.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2041976 | NM_009651.3 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2041976 | NM_001042542.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P212509 | U07423.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P212509 | U10341.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P212509 | BC130271.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P212509 | NM_009651.3 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P212509 | NM_001042542.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P157795 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P110585 | - | - |
Whole Mouse Genome G4122A | A_51_P212509 | U07423.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P212509 | U10341.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P212509 | BC130271.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P212509 | NM_009651.3 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P212509 | NM_001042542.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P106513 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P157795 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P110585 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32236854 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32236722 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32236724 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32236774 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32236869 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32236767 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32236795 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32236853 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32236733 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32236796 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32236743 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32236848 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32236804 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32236820 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32236840 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32236837 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32236841 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32236860 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32236704 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32236814 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32236712 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32236791 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32236801 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32236792 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32236766 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32236818 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32236842 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32236730 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32236863 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32236765 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32236806 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32236771 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32236717 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32236862 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32236855 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32236829 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32236831 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32236788 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32236707 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32236845 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32236720 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32236701 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32236836 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32236783 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32236793 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32236777 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32236819 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32236726 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32236702 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32236736 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32236759 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32236784 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32236746 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32236740 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32236715 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32236775 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32236803 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32236768 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32236802 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32236809 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32236798 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32236835 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32236832 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32236725 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32236852 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32236780 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32236821 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32236735 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32236861 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32236867 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32236782 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32236834 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32236731 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32236700 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32236761 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32236744 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32236779 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32236800 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32236844 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32236781 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32236699 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32236850 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32236769 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32236826 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32236857 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32236711 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32236849 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32236797 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32236779 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32236854 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32236759 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32236781 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32236826 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32236801 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32236761 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32236818 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32236767 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32236720 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32236725 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32236743 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32236712 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32236729 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32236722 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32236735 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32236715 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P15107766 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14722746 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32236726 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32236744 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14532228 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32236724 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32236730 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14554910 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P15107759 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32236775 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32236856 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14532236 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14983080 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32236740 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32236739 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32236868 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32236774 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32236733 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32236780 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32236857 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32236867 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P15107757 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32236783 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P15196491 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P15107764 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32236784 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P15215802 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32236821 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14722747 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32236862 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32236777 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32236800 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32236861 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32236823 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32236769 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32236707 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32236746 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32236770 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32236803 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32236748 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32236782 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32236804 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32236768 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32236863 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32236731 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32236766 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32236809 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32236709 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32236802 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32236824 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P15196488 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32236736 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32236765 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P32236711 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10598280 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10598285 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10598283 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10598282 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10598281 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10598286 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10598284 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1451662_PM_x_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1419019_PM_a_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1451661_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10598279 | - | - |
Mouse Genome 430 2.0 Array | 1451662_x_at | - | - |
Mouse Genome 430 2.0 Array | 1451661_at | - | - |
Mouse Genome 430 2.0 Array | 1431868_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1419019_PM_a_at | - | - |
Mouse Genome 430 2.0 Array | 1451662_PM_x_at | - | - |
Mouse Genome 430 2.0 Array | 1419019_a_at | - | - |
Mouse Genome 430 2.0 Array | 1431868_at | - | - |
Mouse Genome 430 2.0 Array | 1451661_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 11643_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 11643_PM_at | - | - |
Affymetrix Murine Genome 6500A Array | Msa13490_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0X00000167.mm.1 | NM_001042542.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0X00000167.mm.1 | TC0X00000054.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0X00000172.mm.1 | NM_001042542.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0X00000172.mm.1 | NM_009651.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0X00000172.mm.1 | TC0X00000054.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0X00000173.mm.1 | NM_001042542.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0X00000173.mm.1 | NM_009651.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0X00000173.mm.1 | TC0X00000054.mm.1 | MTA TC |
Mouse Expression Set 430 | 1451662_x_at | - | - |
Mouse Expression Set 430 | 1451661_PM_at | - | - |
Mouse Expression Set 430 | 1419019_a_at | - | - |
Mouse Expression Set 430 | 1451661_at | - | - |
Mouse Expression Set 430 | 1419019_PM_a_at | - | - |
Mouse Expression Set 430 | 1451662_PM_x_at | - | - |
Murine Genome U74v2 Set | 102857_at | - | - |
Murine Genome U74v2 Set | 102857_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1451662_x_at | - | - |
Mouse Genome 430A 2.0 Array | 1451661_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1419019_PM_a_at | - | - |
Mouse Genome 430A 2.0 Array | 1451662_PM_x_at | - | - |
Mouse Genome 430A 2.0 Array | 1451661_at | - | - |
Mouse Genome 430A 2.0 Array | 1419019_a_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1451662_x_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1419019_PM_a_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1419019_a_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1451661_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1451662_PM_x_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1451661_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 824156 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 3520619 | - | - |
MouseWG-6 v2 Expression BeadChip | 2190731 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 3520619 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 2190731 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 104070204 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0003520619 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0002190731 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2773339 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2648635 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2773339 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2759971 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2648635 | - | - |
mouse-6 v1.1 (Illumina) | scl0002921.1_68-S | - | - |
mouse-6 v1.1 (Illumina) | scl0002930.1_6-S | - | - |
mouse-6 v1.1 (Illumina) | scl55085.7.1_52-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 3520619 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 2190731 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 104070204 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl0002930.1_6-S | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl55085.7.1_52-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 104070204 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2190731 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3520619 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_3128227 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2648635 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2773339 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_3051805 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2759971 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2648635 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3128227 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2773339 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3051805 | - | - |
