Assay Details
Gene Symbol: Net1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047159.2 | NP_001040624.1 | 3 - 4 | 564 | 98 | |
NM_019671.3 | NP_062645.2 | 5 - 6 | 685 | 98 | ||
GenBank mRNA | AF094520.1 | - | 5 - 6 | 581 | 98 | |
AJ010045.1 | - | 3 - 4 | 527 | 98 | ||
AK082128.1 | - | 3 - 4 | 561 | 98 | ||
AK088632.1 | - | 3 - 4 | 528 | 98 | ||
AK139921.1 | - | 3 - 4 | 528 | 98 | ||
BC004699.1 | - | 3 - 4 | 538 | 3499258 | 98 | |
CN702942.1 | - | 3 - 4 | 425 | 98 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
