Assay Details
Gene Symbol: Hist1h4k | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178211.2 | NP_835583.1 | 1 | 199 | 138 | |
GenBank mRNA | AK139521.1 | - | 1 | 186 | 138 | |
BC096767.1 | - | 166 | 138 | |||
BC117010.1 | - | 1 | 199 | 40126329 | 138 | |
BC117012.1 | - | 1 | 199 | 40126331 | 138 | |
BC119612.1 | - | 1 | 151 | 40061668 | 138 |
Gene Symbol: Hist1h4f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC115446.1 | - | 1 | 144 | 138 |
Gene Symbol: Hist1h4m | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195421.1 | NP_001182350.1 | 1 | 189 | 138 |
Gene Symbol: Hist1h4n | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175657.2 | NP_783588.1 | 1 | 151 | 138 | |
GenBank mRNA | BC115448.2 | - | 1 | 151 | 40099764 | 138 |
BC152397.1 | - | 1 | 151 | 40099768 | 138 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28874404 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2091145 | BC096767.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2091145 | BC117010.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2091145 | BC117012.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2091145 | NM_178211.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2091145 | AK139521.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2091147 | BC096767.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2091147 | BC117010.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2091147 | BC117012.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2091147 | BC119612.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2091147 | NM_178211.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 86825 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 776487 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2091145 | BC096767.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2091145 | BC117010.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2091145 | BC117012.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2091145 | NM_178211.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2091145 | AK139521.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2091147 | BC096767.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2091147 | BC117010.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2091147 | BC117012.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2091147 | BC119612.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2091147 | NM_178211.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28874419 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28874396 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28874393 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28874399 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28874428 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28874392 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28874394 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28874413 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28874425 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28874390 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28874415 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28874397 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28874429 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28874404 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28874401 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28874423 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28874416 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28874400 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28874414 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28874395 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P28874421 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P02644498 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P02644498 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10893019 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10918096 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P10918097 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P28874404 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874404 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874374 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874428 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874477 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874446 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874425 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874470 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874389 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874442 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874383 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874423 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874394 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874449 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874468 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874479 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874395 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874400 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874460 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874435 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874396 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874436 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874385 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874413 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874390 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874455 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874459 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874463 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874421 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874475 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874452 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874419 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874466 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874447 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874379 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874380 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874401 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874397 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874387 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874456 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874392 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874399 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874375 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P28874445 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28874385 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28874419 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28874401 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28874389 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28874379 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28874380 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28874383 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28874425 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28874421 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28874390 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28874428 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28874396 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28874413 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28874423 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10880985 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28874387 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P10474273 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28874404 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28874375 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28874392 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28874377 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28874394 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28874411 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28874415 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28874370 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28874374 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28874416 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28874395 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28874400 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28874399 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P28874397 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10408075 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10408076 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10408074 | - | - |
Affymetrix Mouse Clariom S Array | PSR1300001204.mm.1 | TC1300000182.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1300001204.mm.1 | NM_001195421.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1300001204.mm.1 | NM_001195421.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1300001204.mm.1 | NM_175657.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1300001203.mm.1 | TC1300000182.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1300001203.mm.1 | NM_001195421.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1300001203.mm.1 | NM_001195421.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1300009832.mm.1 | TC1300001557.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1300009833.mm.1 | TC1300001557.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1300009833.mm.1 | NM_178211.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1300009834.mm.1 | TC1300001557.mm.1 | MTA TC |
Mouse Expression Set 430 | 1422948_PM_s_at | - | - |
Mouse Expression Set 430 | 1422948_s_at | - | - |
Mouse Expression Set 430 | 1428014_at | - | - |
Mouse Expression Set 430 | 1424854_PM_at | - | - |
Mouse Expression Set 430 | 1428014_PM_at | - | - |
Mouse Expression Set 430 | 1424854_at | - | - |
Mouse Expression Set 430 | 1422947_at | - | - |
Mouse Expression Set 430 | 1422947_PM_at | - | - |
Mouse Expression Set 430 | 1431615_at | - | - |
Mouse Expression Set 430 | 1431658_at | - | - |
Mouse Expression Set 430 | 1431615_PM_at | - | - |
Mouse Expression Set 430 | 1431658_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1422947_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1422947_at | - | - |
Mouse Genome 430A 2.0 Array | 1428014_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1428014_at | - | - |
Mouse Genome 430A 2.0 Array | 1422948_PM_s_at | - | - |
Mouse Genome 430A 2.0 Array | 1422948_s_at | - | - |
Mouse Genome 430A 2.0 Array | 1424854_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1424854_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 103850348 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 103850348 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 4570735 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0004570735 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1243654 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1243654 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2525570 | - | - |
mouse-6 v1.1 (Illumina) | GI_30061358-S | - | - |
mouse-6 v1.1 (Illumina) | scl0319160.1_25-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 4570735 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 103850348 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl0319160.1_25-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 103850348 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4570735 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1243654 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1243654 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2525570 | - | - |
