Assay Details
Gene Symbol: Cdc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025779.1 | NP_001020950.1 | 6 - 7 | 1050 | 90 | |
NM_011799.2 | NP_035929.1 | 6 - 7 | 1126 | 90 | ||
GenBank mRNA | AJ009559.1 | - | 6 - 7 | 1143 | 90 | |
AJ223087.1 | - | 6 - 7 | 1128 | 90 | ||
AK012192.1 | - | 6 - 7 | 1080 | 90 | ||
AK075975.1 | - | 6 - 7 | 1078 | 90 | ||
AK085020.1 | - | 6 - 7 | 1080 | 90 | ||
AK162669.1 | - | 6 - 7 | 1088 | 90 | ||
AK166003.1 | - | 6 - 7 | 1131 | 90 | ||
BC052434.1 | - | 6 - 7 | 1050 | 6837113 | 90 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
