Assay Details
Gene Symbol: Mycl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK146959.1 | - | 3 | 3029 | 152 | |
BC037764.1 | - | 1 | 807 | 152 | ||
BC053059.1 | - | 2 | 3368 | 5704829 | 152 | |
BC089346.1 | - | 3 | 3004 | 6810905 | 152 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23260203 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23260194 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23260191 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23260198 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23260200 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23260204 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23260201 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23260193 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23260205 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23260192 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23260196 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23260195 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23260199 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23260197 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1972025 | BC089346.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1972025 | BC053059.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1972025 | BC037764.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1972025 | NM_008506.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P1972025 | AK146959.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 494107 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 593873 | AK090039.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 495154 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P135652 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1972025 | BC089346.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1972025 | BC053059.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1972025 | BC037764.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1972025 | NM_008506.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1972025 | AK146959.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23260179 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23260201 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23260199 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23260193 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23260215 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23260177 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23260194 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23260187 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23260198 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23260185 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23260210 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23260206 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23260208 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23260186 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23260172 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23260203 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23260205 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23260207 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23260184 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23260189 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23260197 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23260174 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23260213 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23260209 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23260180 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P23260183 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P135652 | - | - |
Whole Mouse Genome G4122A | A_52_P93284 | BC089346.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P93284 | BC053059.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P93284 | BC037764.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P93284 | NM_008506.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P93284 | AK146959.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P109761 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P127397 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P135652 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03534901 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03534903 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03534902 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03534896 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23260203 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23260201 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23260192 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23260200 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23260191 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23260194 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23260195 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23260204 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23260199 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23260196 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23260205 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23260198 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23260197 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P23260193 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23260185 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23260248 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23260164 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23260262 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23260228 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23260246 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23260203 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23260213 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23260243 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23260271 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23260258 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23260261 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23260245 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23260291 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23260217 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23260194 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23260270 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23260227 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23260240 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23260257 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23260208 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23260224 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23260186 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23260209 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23260267 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23260199 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23260177 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23260193 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23260215 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23260226 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23260205 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23260219 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23260266 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23260221 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23260159 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23260286 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23260233 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23260168 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23260189 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23260273 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23260275 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23260183 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23260229 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23260231 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23260207 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23260253 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23260210 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23260256 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23260197 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23260201 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23260161 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23260175 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23260222 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23260235 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23260187 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23260237 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P23260278 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23260224 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23260267 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23260245 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23260219 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23260234 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23260264 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23260215 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23260217 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03882437 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23260253 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23260266 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03848589 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03882440 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23260176 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03534894 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04086440 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23260222 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23260231 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23260158 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03534902 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23260208 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23260240 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03704099 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23260213 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23260161 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23260235 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23260270 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03554190 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23260265 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23260271 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23260193 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04018193 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23260169 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04021741 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23260221 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03554183 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03882434 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03821884 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23260261 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23260256 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23260163 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23260243 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23260166 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23260175 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23260254 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23260179 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04058619 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23260197 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23260209 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23260183 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23260187 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23260168 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23260227 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23260233 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23260171 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23260189 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23260185 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23260251 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23260273 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23260194 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23260205 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04018192 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23260177 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04018189 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23260159 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23260226 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23260201 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23260248 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10507803 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10507800 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10507799 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10507801 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1422087_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1422088_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1434777_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10507798 | - | - |
Mouse Genome 430 2.0 Array | 1434777_at | - | - |
Mouse Genome 430 2.0 Array | 1434777_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1422088_at | - | - |
Mouse Genome 430 2.0 Array | 1422087_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1422088_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1422087_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 16918_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 16918_PM_at | - | - |
Mouse Expression Set 430 | 1422087_PM_at | - | - |
Mouse Expression Set 430 | 1434777_PM_at | - | - |
Mouse Expression Set 430 | 1422088_PM_at | - | - |
Mouse Expression Set 430 | 1422088_at | - | - |
Mouse Expression Set 430 | 1434777_at | - | - |
Mouse Expression Set 430 | 1422087_at | - | - |
Murine Genome U74v2 Set | 102235_at | - | - |
Murine Genome U74v2 Set | 102235_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1422087_at | - | - |
Mouse Genome 430A 2.0 Array | 1422088_at | - | - |
Mouse Genome 430A 2.0 Array | 1434777_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1422087_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1422088_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1434777_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1422087_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1422088_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1434777_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1422088_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1434777_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1422087_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 1980309 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 1980309 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0001980309 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1221750 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1221750 | - | - |
mouse-6 v1.1 (Illumina) | scl24999.4_423-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 1980309 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl24999.4_423-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1980309 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1221750 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1221750 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl24999.4_423-S | - | - |
