Assay Details
Gene Symbol: A930012L18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_026853.1 | 2 - 3 | 720 | 80 | ||
GenBank mRNA | AK044430.1 | - | 2 - 3 | 724 | 80 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | mutated in colorectal cancers | Mcc |
D18Ertd451e, E330037C19 |
Anti-sense | Chr. 18 - 44425060 - 44812182 on NCBI Build GRCm38 | |
Downstream | YTH domain containing 2 | Ythdc2 |
3010002F02Rik, BC037178 |
155.4kb | Sense | Chr. 18 - 44827744 - 44889720 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
